Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bai
- Entrez Gene ID
-
42996
- Gene Name
- baiser
- Gene Aliases
-
150547_at, BAI, Bai, BcDNA:RE52411, CG11785, Dmel\CG11785, Dmel_CG11785, p23, p24
- Chromosome Location
-
Chr.3R: 25039374 - 25040443 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045866
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143066.3 |
NP_651323.3 |
3 - 4 |
450 |
85 |
GenBank mRNA |
AY075503.1 |
- |
3 - 4 |
449 |
85 |
View Full Product Details