Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TwdlS
Entrez Gene ID
43209
Gene Name
TweedleS
Gene Aliases
CG14242, Dmel\CG14242, Dmel_CG14242
Chromosome Location
Chr.3R: 26636688 - 26637557 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039443
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143234.2 NP_651491.1 1 - 2 70 74
GenBank mRNA BT149868.1 - 1 - 2 44 74

View Full Product Details