Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gb
Entrez Gene ID
43265
Gene Name
genderblind
Gene Aliases
CG6070, Dmel\CG6070, Dmel_CG6070, Gb, anon-WO0118547.295
Chromosome Location
Chr.3R: 27108467 - 27112270 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039487
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300626.1 NP_001287555.1 4 - 5 1419 57
NM_001316513.1 NP_001303442.1 4 - 5 1864 57
NM_143279.3 NP_651536.1 4 - 5 1864 57
GenBank mRNA AY060663.1 - 4 - 5 1827 57
BT011041.1 - - 3119 57

View Full Product Details