Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PH4alphaNE3
Entrez Gene ID
326114
Gene Name
prolyl-4-hydroxylase-alpha NE3
Gene Aliases
CG31017, CG9708, Dmel\CG31017, Dmel_CG31017
Chromosome Location
Chr.3R: 30604181 - 30606144 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051017
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143557.2 NP_651814.1 3 - 4 470 90
GenBank mRNA AF495538.1 - 3 - 4 470 90
AY051566.1 - 3 - 4 470 90

View Full Product Details