Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sle
Entrez Gene ID
41262
Gene Name
slender lobes
Gene Aliases
CG12819, CG12819-PA, Dmel\CG12819, Dmel_CG12819, FBtr0082231, SLE, Sle
Chromosome Location
Chr.3R: 10333115 - 10338265 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037810
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169338.2 NP_731472.1 3 - 4 3800 70
NM_169339.2 NP_731473.1 2 - 3 3701 70
GenBank mRNA BT031151.1 - 3 - 4 3739 70

View Full Product Details