Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpr15
Entrez Gene ID
41473
Gene Name
defective proboscis extension response 15
Gene Aliases
CG10095, CT28423, Dmel\CG10095, Dmel_CG10095, Dpr-15, Dpr15
Chromosome Location
Chr.3R: 12138833 - 12158595 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037993
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300370.1 NP_001287299.1 4 - 5 1369 64
NM_169453.1 NP_731672.1 4 - 5 1385 64
GenBank mRNA BT044240.1 - 4 - 5 1385 64

View Full Product Details