Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cad88C
- Entrez Gene ID
-
41774
- Gene Name
- Cadherin 88C
- Gene Aliases
-
CG3389, CT11389, DCad88C, Dm Cad88C, Dmel\CG3389, Dmel_CG3389, cad88C
- Chromosome Location
-
Chr.3R: 14630478 - 14641776 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038247
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169594.3 |
NP_731930.3 |
11 - 12 |
6379 |
76 |
View Full Product Details