Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dscam3
Entrez Gene ID
42103
Gene Name
Down syndrome cell adhesion molecule 3
Gene Aliases
CG12536, CG31190, CG8106, CT33960, Dmel\CG31190, Dmel_CG31190, Dscam-like
Chromosome Location
Chr.3R: 17473935 - 17505667 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261046
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104355.2 NP_001097825.1 24 - 25 4831 67
NM_169761.2 NP_732242.2 24 - 25 4354 67
NM_206504.2 NP_996226.2 24 - 25 4354 67

View Full Product Details