Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Takl1
- Entrez Gene ID
-
318725
- Gene Name
- Tak1-like 1
- Gene Aliases
-
CG31421, Dmel\CG31421, Dmel_CG31421, FBgn0046689, TAKL1, anon-AE003732.1, takl1
- Chromosome Location
-
Chr.3R: 20770828 - 20772601 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046689
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169924.3 |
NP_732554.1 |
3 - 4 |
699 |
69 |
GenBank mRNA |
BT022955.1 |
- |
3 - 4 |
668 |
69 |
View Full Product Details