Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syp
Entrez Gene ID
42460
Gene Name
Syncrip
Gene Aliases
AI945337, CG17838, Dmel\CG17838, Dmel_CG17838, anon-WO0118547.613, cg17838, l(3)03806, syp
Chromosome Location
Chr.3R: 20756729 - 20810726 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038826
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275847.1 NP_001262776.1 12 - 13 2285 67
NM_001275850.1 NP_001262779.1 13 - 14 2569 67
NM_001275851.1 NP_001262780.1 12 - 13 3894 67
NM_001275852.1 NP_001262781.1 11 - 12 2024 67
NM_001275853.1 NP_001262782.1 12 - 13 2559 67
NM_001275855.1 NP_001262784.1 11 - 12 3610 67
NM_001369986.1 NP_001356920.1 13 - 14 2569 67
NM_169926.2 NP_732556.1 12 - 13 2308 67

View Full Product Details