Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Wsck
- Entrez Gene ID
-
318600
- Gene Name
- CG31127-PA
- Gene Aliases
-
BcDNA:LD25626, BcDNA:SD05152, CG31127, Dmel\CG31127, Dmel_CG31127, WSCK
- Chromosome Location
-
Chr.3R: 24627066 - 24630075 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046685
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170155.2 |
NP_733018.1 |
2 - 3 |
1302 |
70 |
GenBank mRNA |
BT001869.1 |
- |
2 - 3 |
1317 |
70 |
View Full Product Details