Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Elal
- Entrez Gene ID
-
42970
- Gene Name
- Elastin-like
- Gene Aliases
-
CG7021, Dmel\CG7021, Dmel_CG7021, Ela
- Chromosome Location
-
Chr.3R: 24865350 - 24867651 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0013949
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170176.2 |
NP_733055.1 |
1 - 2 |
313 |
54 |
View Full Product Details