Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
faf
Entrez Gene ID
43749
Gene Name
fat facets
Gene Aliases
BcDNA.LD22582, BcDNA:LD22582, CG1945, Dmel\CG1945, Dmel_CG1945, Faf, Fat, P55824, USPX
Chromosome Location
Chr.3R: 31777020 - 31790681 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005632
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260458.1 NP_001247387.1 15 - 16 7567 68
NM_001260459.1 NP_001247388.1 15 - 16 7615 68
NM_001260460.2 NP_001247389.2 15 - 16 7583 68
NM_001260461.1 NP_001247390.1 16 - 17 7681 68
NM_001300698.1 NP_001287627.1 15 - 16 7583 68
NM_079873.5 NP_524612.2 15 - 16 7583 68
NM_170576.3 NP_733455.1 15 - 16 7583 68

View Full Product Details