Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
faf
Entrez Gene ID
43749
Gene Name
fat facets
Gene Aliases
BcDNA.LD22582, BcDNA:LD22582, CG1945, Dmel\CG1945, Dmel_CG1945, Faf, Fat, P55824, USPX
Chromosome Location
Chr.3R: 31777020 - 31790681 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005632
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260458.1 NP_001247387.1 17 - 18 8543 81
NM_001260461.1 NP_001247390.1 18 - 19 8657 81
NM_001300698.1 NP_001287627.1 17 - 18 8559 81
NM_079873.5 NP_524612.2 17 - 18 8574 81

View Full Product Details