Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bon
Entrez Gene ID
44235
Gene Name
bonus
Gene Aliases
0241/08, 0249/12, 0434/20, 0487/06, 150268_at, 150269_at, Bon, Bonus, CG15687, CG5206, Dmel\CG5206, Dmel_CG5206, anon-WO0118547.418, l(3)S043420
Chromosome Location
Chr.3R: 20593139 - 20612540 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023097
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170194.2 NP_001163665.1 5 - 6 2885 74
NM_001275836.1 NP_001262765.1 5 - 6 2909 74
NM_001275837.1 NP_001262766.1 5 - 6 2891 74
NM_001275838.1 NP_001262767.1 5 - 6 2909 74
NM_001275839.1 NP_001262768.1 5 - 6 2903 74
NM_079985.4 NP_524724.2 5 - 6 2909 74
GenBank mRNA BT031160.1 - 5 - 6 2865 74

View Full Product Details