Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stops
Entrez Gene ID
43683
Gene Name
slow termination of phototransduction
Gene Aliases
CG15557, CG15558, CG15559, CG31006, Dmel\CG31006, Dmel_CG31006, su(rdgB)69
Chromosome Location
Chr.3R: 31158156 - 31169106 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086704
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170309.2 NP_001163780.1 5 - 6 1075 69
NM_170537.2 NP_733416.2 4 - 5 891 69
NM_170538.3 NP_733417.1 5 - 6 1030 69
GenBank mRNA AY047558.1 - 4 - 5 647 69

View Full Product Details