Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mical
Entrez Gene ID
41225
Gene Name
Molecule interacting with CasL
Gene Aliases
CG11685, CG11687, CG18484, CG18667, CG18668, CG33186, CG33190, CG33208, Dmel\CG33208, Dmel_CG33208, MICAL, mdcds_49226, mical, milch
Chromosome Location
Chr.3R: 10001554 - 10042895 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053208
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260085.2 NP_001247014.1 20 - 21 14078 67
NM_001260086.3 NP_001247015.1 21 - 22 14159 67
NM_001260087.3 NP_001247016.2 21 - 22 14399 67
NM_001316573.1 NP_001303502.1 19 - 20 14024 67
NM_001316574.1 NP_001303503.1 18 - 19 8930 67
NM_176443.2 NP_788620.1 17 - 18 8159 67
NM_176444.2 NP_788621.1 19 - 20 8963 67
NM_176445.2 NP_788622.1 18 - 19 14004 67
NM_176446.2 NP_788623.1 17 - 18 13960 67
NM_176447.2 NP_788624.1 21 - 22 14126 67
NM_176448.2 NP_788625.1 18 - 19 13900 67
NM_176449.3 NP_788626.2 17 - 18 13844 67

View Full Product Details