Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pxb
Entrez Gene ID
41899
Gene Name
CG33207-PB
Gene Aliases
BcDNA:RE16319, CG14873, CG14874, CG33207, CG33207/Pxb, Dmel\CG33207, Dmel_CG33207, Pxb
Chromosome Location
Chr.3R: 15666183 - 15690513 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053207
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275680.1 NP_001262609.1 3 - 4 940 60
NM_176504.2 NP_788681.1 3 - 4 940 60
GenBank mRNA AB081475.1 - 3 - 4 822 60
BT126212.1 - 3 - 4 917 60

View Full Product Details