Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr97a
- Entrez Gene ID
-
117338
- Gene Name
- Gustatory receptor 97a
- Gene Aliases
-
97D.1, 97a, CG31073, CG31074, CG33083, Dmel\CG33083, Dmel_CG33083, GR97D.1, GR97a, Gr97D1
- Chromosome Location
-
Chr.3R: 26965124 - 26966485 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041224
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176570.2 |
NP_788747.1 |
1 - 2 |
1054 |
67 |
View Full Product Details