Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpr9
Entrez Gene ID
2768670
Gene Name
defective proboscis extension response 9
Gene Aliases
BcDNA:GH01517, CG12601, CG14862, CG14863, CG31114, CG33485, CT34680, Dmel\CG33485, Dmel_CG33485, Dpr-9, Dpr9
Chromosome Location
Chr.3R: 15054930 - 15077206 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038282
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300403.1 NP_001287332.1 5 - 6 1842 66
NM_206493.2 NP_996215.1 5 - 6 1842 66
GenBank mRNA AY102651.1 - 5 - 6 1824 66

View Full Product Details