Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- yip7
- Entrez Gene ID
-
38680
- Gene Name
- yippee interacting protein 7
- Gene Aliases
-
CG6457, Dmel\CG6457, Dmel_CG6457, MAC, SP116, SPH116, Yip7, mac
- Chromosome Location
-
Chr.3L: 6035216 - 6036175 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040060
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079220.2 |
NP_523944.1 |
1 - 2 |
289 |
150 |
GenBank mRNA |
AF512510.1 |
- |
1 - 2 |
275 |
150 |
View Full Product Details