Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sun
Entrez Gene ID
44046
Gene Name
stunted
Gene Aliases
ATP5E, ATPE, ATPsynepsilon, BcDNA:RE19513, BcDNA:RH48911, CG9032, Dmel\CG9032, Dmel_CG9032, Sun, l(1)13Fe, l(1)XV
Chromosome Location
Chr.X: 15847859 - 15849258 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014391
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079943.4 NP_524682.1 2 - 3 260 130
GenBank mRNA AY071158.1 - 2 - 3 242 130
AY071744.1 - 2 - 3 243 130

View Full Product Details