Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Orc6
- Entrez Gene ID
-
36017
- Gene Name
- Origin recognition complex subunit 6
- Gene Aliases
-
CG1584, DmORC 6, DmORC6, Dmel\CG1584, Dmel_CG1584, ORC, ORC6, orc6, rDmORC
- Chromosome Location
-
Chr.2R: 9845458 - 9846485 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0023180
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057971.6 |
NP_477319.1 |
2 - 2 |
790 |
89 |
GenBank mRNA |
AY071443.1 |
- |
2 - 2 |
765 |
89 |
View Full Product Details