Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krn
Entrez Gene ID
326198
Gene Name
Keren
Gene Aliases
CG32179, CG8056, CT19073, Dmel\CG32179, Dmel_CG32179, KRN, Ker, gritz, keren, keren/spitz2, krn, s-krn, skrn
Chromosome Location
Chr.3L: 17649562 - 17654280 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052179
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079405.4 NP_524129.1 2 - 2 1045 139
GenBank mRNA AF245387.1 - 2 - 2 1013 139
AY060409.1 - 2 - 2 1013 139

View Full Product Details