Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TotZ
- Entrez Gene ID
-
117459
- Gene Name
- Turandot Z
- Gene Aliases
-
CG31507, Dmel\CG31507, Dmel_CG31507, Totz, tot
- Chromosome Location
-
Chr.3R: 20877736 - 20878363 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0044809
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080521.3 |
NP_536782.2 |
2 - 2 |
434 |
152 |
View Full Product Details