Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAN1
Entrez Gene ID
37838
Gene Name
CG3167-PA
Gene Aliases
BcDNA:LD24895, CG3167, Dmel\CG3167, Dmel_CG3167, dMAN1, dMan1
Chromosome Location
Chr.2R: 24033974 - 24036707 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034962
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299883.1 NP_001286812.1 2 - 2 2292 50
NM_138029.3 NP_611873.1 2 - 2 2029 50
NM_166651.2 NP_726404.1 2 - 2 2041 50
GenBank mRNA BT001698.1 - 2 - 2 2057 50

View Full Product Details