Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kuk
Entrez Gene ID
42063
Gene Name
kugelkern
Gene Aliases
CG5175, Dmel\CG5175, Dmel_CG5175, Kuk, char, kuk/char
Chromosome Location
Chr.3R: 17080244 - 17084781 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038476
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260219.1 NP_001247148.1 2 - 2 1728 83
NM_142338.3 NP_650595.1 2 - 2 1759 83
NM_169745.3 NP_732204.1 2 - 2 1766 83
GenBank mRNA AY094778.1 - 2 - 2 1770 83

View Full Product Details