Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vkor
Entrez Gene ID
3346188
Gene Name
Vitamin-K epoxide reductase
Gene Aliases
BP1071, CG33544, Dmel\CG33544, Dmel_CG33544, HDC06808, VKORC1
Chromosome Location
Chr.2R: 16778365 - 16778994 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053544
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014533.2 NP_001014533.1 1 - 2 170 86

View Full Product Details