Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Crg-1
- Entrez Gene ID
-
43900
- Gene Name
- Circadianly Regulated Gene
- Gene Aliases
-
CG32788, Crg1, Dmel\CG32788, Dmel_CG32788, crg1
- Chromosome Location
-
Chr.X: 3793223 - 3797339 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0021738
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166990.4 |
NP_726887.1 |
3 - 3 |
854 |
149 |
View Full Product Details