Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Bem46
- Entrez Gene ID
-
44441
- Gene Name
- Bem46-PA
- Gene Aliases
-
CG18642, Dmel\CG18642, Dmel_CG18642, c1.6
- Chromosome Location
-
Chr.2L: 2988116 - 2990075 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0025109
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058024.4 |
NP_477372.1 |
3 - 4 |
857 |
83 |
GenBank mRNA |
AF069781.1 |
- |
3 - 4 |
803 |
83 |
- Gene Symbol
- mod(mdg4)
- Entrez Gene ID
-
49228
- Gene Name
- modifier of mdg4
- Gene Aliases
-
BcDNA:GH07769, BcDNA:SD03001, CG15500, CG15501, CG15802, CG18151, CG32491, CG7836, CG7859, CG8076, Dmel\CG32491, Dmel_CG32491, Doom, E(var)129, E(var)3-93D, E(var)93D, E-(var)3-93D, E-var(3)1, E-var(3)3, LBC, MOD(MDG4), MOD(MDG4)56.3, MOD2.2, Mod(mdg 4), Mod(mdg 4)67.2, Mod(mdg)4, Mod(mdg4), Mod(mdg4)-67.2, Mod(mdg4)2.2, Mod(mdg4)67.2, Mod2.2, Mod67.2, Mod[mdg4], R5_3R:17176815..17177103, bpd, doom, l(3)03852, l(3)L3101, l(3)j2B7, mdg4, mmd4, mnm, mnn, mod, mod (mdg4), mod(gypsy), mod(mdg)4, mod(mdg-4), mod(mdg4, mod(mdg4)-58.8, mod(mdg4)-67.2, mod(mdg4)67.2, mod-mdg4, mod2.2, modMDG4, mod[mdg4], pf-2
- Chromosome Location
-
Chr.3R: 21370101 - 21377401 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002781
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BT003484.1 |
- |
- |
777 |
83 |
View Full Product Details