Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ORY
Entrez Gene ID
3355174
Gene Name
Occludin-Related Y
Gene Aliases
CG40446, Dmel\CG40446, Dmel_CG40446, Ory, ory
Chromosome Location
Chr.Y: 3561745 - 3618339 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0046323
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015498.5 NP_001015498.5 6 - 7 2277 137

View Full Product Details