Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jet
Entrez Gene ID
33705
Gene Name
jetlag
Gene Aliases
CG8873, Dmel\CG8873, Dmel_CG8873, FBXL15, JET, JETLAG, Jet
Chromosome Location
Chr.2L: 4948206 - 4949784 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031652
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042867.2 NP_001036332.1 1 - 2 890 74

View Full Product Details