Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cep43
Entrez Gene ID
415215
Gene Name
centrosomal protein 43
Gene Aliases
ccr6a, fgfr1op, zgc:86782
Chromosome Location
Chr.20: 30578966 - 30610172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002125.1 NP_001002125.1 4 - 5 301 78
XM_005160662.4 XP_005160719.1 4 - 5 335 78
XM_005160663.4 XP_005160720.1 4 - 5 335 78
XM_005160664.4 XP_005160721.1 4 - 5 335 78
XM_017352461.2 XP_017207950.1 4 - 5 335 78
XM_021468390.1 XP_021324065.1 4 - 5 335 78
XM_021468391.1 XP_021324066.1 4 - 5 335 78
XM_021468392.1 XP_021324067.1 4 - 5 335 78
XM_021468393.1 XP_021324068.1 4 - 5 335 78
GenBank mRNA BC071450.1 - 4 - 5 301 78

View Full Product Details