Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nfs1
Entrez Gene ID
562714
Gene Name
NFS1 cysteine desulfurase
Gene Aliases
fb50g03, wu:fb50g03
Chromosome Location
Chr.11: 24348425 - 24356065 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044993.1 NP_001038458.1 5 - 6 722 111
GenBank mRNA BC134974.1 - 5 - 6 719 111

View Full Product Details