Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrps18c
Entrez Gene ID
569841
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
si:ch211-85e10.6
Chromosome Location
Chr.21: 19328987 - 19330302 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045190.2 NP_001038655.2 1 - 2 122 111
GenBank mRNA BC134080.1 - 1 - 2 122 111

View Full Product Details