Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tyrobp
Entrez Gene ID
100101461
Gene Name
transmembrane immune signaling adaptor TYROBP
Gene Aliases
dap12, tyrpbp
Chromosome Location
Chr.16: 25195602 - 25200757 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100103.2 NP_001093573.2 2 - 3 134 64
GenBank mRNA EF158446.1 - 2 - 3 134 64

View Full Product Details