Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- spon2b
- Entrez Gene ID
-
30202
- Gene Name
- spondin 2b, extracellular matrix protein
- Gene Aliases
-
etID309958.14, mindin2, zgc:100756
- Chromosome Location
-
Chr.14: 17376968 - 17382258 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB006085.1 |
- |
1 - 2 |
875 |
91 |
View Full Product Details