Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tgif1
Entrez Gene ID
321756
Gene Name
TGFB-induced factor homeobox 1
Gene Aliases
Tgif, fa06h05, fb33g11, wu:fa06h05, wu:fb33g11, zgc:55852
Chromosome Location
Chr.24: 9287412 - 9293918 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199567.1 NP_955861.1 2 - 3 467 61
GenBank mRNA AY391467.1 - 2 - 3 440 61
BC044510.1 - 2 - 3 467 61

View Full Product Details