Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gbgt1l1
Entrez Gene ID
393300
Gene Name
globoside alpha-1,3-N-acetylgalactosaminyltransferase 1, like 1
Gene Aliases
zgc:56701
Chromosome Location
Chr.5: 23702295 - 23715815 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200329.1 NP_956623.1 3 - 4 229 131
GenBank mRNA BC052142.1 - 3 - 4 229 131

View Full Product Details