Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtx3
Entrez Gene ID
402916
Gene Name
metaxin 3
Gene Aliases
zgc:109718
Chromosome Location
Chr.5: 51885258 - 51903002 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205621.1 NP_991184.1 2 - 3 161 74
XM_005165510.4 XP_005165567.3 2 - 3 224 74
XM_017356049.2 XP_017211538.1 2 - 3 228 74
GenBank mRNA AY194226.1 - 2 - 3 161 74
BC094962.1 - 2 - 3 161 74
BC164101.1 - 2 - 3 183 74

View Full Product Details