Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gmppaa
Entrez Gene ID
431743
Gene Name
GDP-mannose pyrophosphorylase Aa
Gene Aliases
zgc:91853
Chromosome Location
Chr.9: 10770612 - 10779267 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002196.1 NP_001002196.1 1 - 2 183 84
XM_005167476.4 XP_005167533.1 1 - 2 314 84
GenBank mRNA BC074036.1 - 1 - 2 183 84

View Full Product Details