Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
surf4l
Entrez Gene ID
436743
Gene Name
surfeit 4, like
Gene Aliases
zgc:92887
Chromosome Location
Chr.5: 63629052 - 63644884 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002470.1 NP_001002470.1 3 - 4 473 70
GenBank mRNA BC076344.1 - 3 - 4 473 70
BC165295.1 - 3 - 4 340 70

View Full Product Details