Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adprh
Entrez Gene ID
445059
Gene Name
ADP-ribosylarginine hydrolase
Gene Aliases
zgc:91926
Chromosome Location
Chr.12: 15148625 - 15196223 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003453.2 NP_001003453.1 5 - 6 725 63
GenBank mRNA BC078399.1 - 5 - 6 571 63
BC164170.1 - 4 - 5 463 63
EH441896.1 - 5 - 6 723 63
EH557169.1 - 5 - 6 766 63

View Full Product Details