Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
illr4
Entrez Gene ID
559737
Gene Name
immune-related, lectin-like receptor 4
Gene Aliases
illrL, zgc:152681
Chromosome Location
Chr.19: 40167363 - 40175776 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040040.1 NP_001035129.1 4 - 5 354 99
GenBank mRNA AY986758.1 - 4 - 5 354 99

View Full Product Details