Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wdyhv1
Entrez Gene ID
560935
Gene Name
WDYHV motif containing 1
Gene Aliases
ntaq1, si:rp71-45k5.3
Chromosome Location
Chr.19: 35002479 - 35004947 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044947.1 NP_001038412.1 3 - 4 232 140
GenBank mRNA BC122321.1 - 2 - 3 167 140
BC171472.1 - 3 - 4 286 140
BC171474.1 - 3 - 4 286 140

View Full Product Details