Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plcd3a
Entrez Gene ID
569040
Gene Name
phospholipase C, delta 3a
Gene Aliases
zgc:158396
Chromosome Location
Chr.3: 39260082 - 39305291 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099423.1 NP_001092893.1 12 - 13 2325 90
GenBank mRNA BC139849.1 - 12 - 13 2325 90

View Full Product Details