The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178306.3 | NP_840091.2 | 9 - 10 | 1199 | 64 |
XM_017354058.2 | XP_017209547.1 | 9 - 10 | 1175 | 64 | |
GenBank mRNA | AF548566.1 | - | 9 - 10 | 1211 | 64 |
BC059550.1 | - | 9 - 10 | 1200 | 64 |