The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194426.3 | NP_919407.2 | 2 - 3 | 687 | 83 |
GenBank mRNA | AF200950.1 | - | 2 - 3 | 573 | 83 |
BC078289.1 | - | 2 - 3 | 587 | 83 |