Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdia4
Entrez Gene ID
554998
Gene Name
protein disulfide isomerase family A, member 4
Gene Aliases
cai, fj23a12, wu:fd20c07, wu:fj23a12, zgc:113965, zgc:136625, zgc:56014, zgc:77773
Chromosome Location
Chr.24: 17334682 - 17341458 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199779.2 NP_956073.2 3 - 4 714 77
GenBank mRNA BC045862.1 - 3 - 4 640 77
BC063979.1 - 3 - 4 725 77
BC117629.1 - 3 - 4 617 77
BC165633.1 - 3 - 4 513 77

View Full Product Details