Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nup43
Entrez Gene ID
405828
Gene Name
nucleoporin 43
Gene Aliases
zgc:77032
Chromosome Location
Chr.20: 1316733 - 1327859 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212892.1 NP_998057.1 1 - 2 183 141
GenBank mRNA BC066616.1 - 1 - 2 183 141

View Full Product Details